Mop frame, aluminium
Leverancier: PFENNIG REINIGUNGSTECHNIK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2009100 2009161
PPSA2009100EA 63.2 EUR
PPSA2009100 PPSA2009161
Mop frame, aluminium
Moppen
Aluminium mop frame with reduced weight.
  • Mechanics made of stainless steel, plate made of aluminium
  • Autoclavable
  • Contactless mop drop with MopDrop