Buckets, Rectangular
Leverancier: PFENNIG REINIGUNGSTECHNIK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3550008 3550009 3550010
PPSA3550008EA 66.5 EUR
PPSA3550008 PPSA3550009 PPSA3550010
Buckets, Rectangular
Emmers
Standard bucket, rectangular, made of PP, with stainless steel handle, for Clino® system trolleys.
  • With scaling 5 to15 L
  • Coloured to facilitate use as part of a colour coded system
  • Good chemical resistance
  • Good temperature resistance
  • Autoclavable (approximately 15 to 30 cycles)