Reversed phase HPLC columns, Synergi™ Hydro-RP
Leverancier: PHENOMENEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
00B-4387-E0 00B-4375-Y0 00B-4375-E0 00G-4375-E0 00B-4387-Y0 00B-4387-B0 00F-4375-E0 00F-4375-Y0 00F-4375-B0 00F-4375-A0 00D-4375-B0 00D-4387-B0 00D-4387-Y0
PHEN00B-4387-E0EA 0 EUR
PHEN00B-4387-E0 PHEN00B-4375-Y0 PHEN00B-4375-E0 PHEN00G-4375-E0 PHEN00B-4387-Y0 PHEN00B-4387-B0 PHEN00F-4375-E0 PHEN00F-4375-Y0 PHEN00F-4375-B0 PHEN00F-4375-A0 PHEN00D-4375-B0 PHEN00D-4387-B0 PHEN00D-4387-Y0
Reversed phase HPLC columns, Synergi™ Hydro-RP
Kolommen voor chromatografie
Synergi™ Hydro-RP columns have polar embedded C18 phase that provides extreme hydrophobic and slight polar selectivity.
  • Ideal for separation of weakly hydrophobic compounds
  • Stable in 100% aqueous mobile phase