Shelving systems for hazardous substances, RWW
Leverancier: asecos
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
27829 27830 27831 27832 27833 27834 27835 27836
MSGA27829EA 719 EUR
MSGA27829 MSGA27830 MSGA27831 MSGA27832 MSGA27833 MSGA27834 MSGA27835 MSGA27836
Shelving systems for hazardous substances, RWW
Meubilair Legborden
Sturdy construction racks with four galvanized steel sumps shelves as storage levels and galvanized rack frames. Ideal for safe and approved storage of water pollutants.
  • Storage levels in 25 mm increments, height-adjustable
  • Load capacity per storage shelf: 200 kg (evenly distributed)
  • Rack system can be extended individually
Certificaten: TÜV approved.
Leveringsinformatie: Flat-pack delivery with assembly instructions.