Tubing, silicone, Versilon® SPX-50
Leverancier: Saint Gobain Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ABX00002 ABX00004 ABX00007 ABX00017 ABX00053 ABX00003
VERNABX00002EA 0 EUR
VERNABX00002 VERNABX00004 VERNABX00007 VERNABX00017 VERNABX00053 VERNABX00003
Tubing, silicone, Versilon® SPX-50
Slangen Transfer Tubing
Silicone, peroxide-cured, translucent.
  • Provides resiliency, long life and durability
  • Ultra-smooth inner bore reduces risk of particulate entrapment
  • Taste and odour free
  • Non-toxic, non-hemolytic and non-pyrogenic
These high-strength silicone tubing produced from a proprietary combination of silicone elastomers, provides durability and versatility in a range of applications. It optimizes critical physical properties such as tensile strength, elongation and compression set, resulting in a more physically durable product. Ideal for veterinary, medical and pharmaceutical applications.
Temperature resistance: up to 204 °C
Shore hardness: 45 ±5
Density: 1,14-1,20
Certificaten: Meets NSF-51 and FDA 21 CFR PART 177.2600 criteria.