No hole circuit breaker lockouts
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
065396 065397 065965 065966
BRDY065396EA 15.02 EUR
BRDY065396 BRDY065397 BRDY065965 BRDY065966
No hole circuit breaker lockouts
Veiligheidssloten
These "No-Hole" breaker lockouts are extremely versatile and easy to install using patented thumbscrew design - no screwdrivers needed. Simply tighten lockout securely onto switch tongue, pull cover over thumbscrew and lock cover to prevent clamp from being loosened.
  • Made of PP and moulded glass-filled nylon
  • Detachable cleats (included) expand the range of applicable breakers
Max. shackle diameter: 7,00 mm
Max. switch width: 16,00 mm
Min. shackle clearance: 20,00 mm