Anti-CD38 Mouse Monoclonal Antibody (PerCP-Cyanine5.5) [clone: HIT2]
Leverancier: Tonbo Biosciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
65-0389-T025 65-0389-T100
65-0389-T025EA 159 EUR
65-0389-T025 65-0389-T100
Anti-CD38 Mouse Monoclonal Antibody (PerCP-Cyanine5.5) [clone: HIT2]
Antilichamen
The HIT2 antibody is specific for human CD38, a 45 kD type II transmembrane glycoprotein expressed on thymocytes, activated T and B cells, and monocytes as well as other non-hematopoietic cells. CD38 is an ectoenzyme that functions to catalyze the synthesis and hydrolysis of cyclic ADP-ribose and is involved in cell signaling, regulating cell adhesion, and activation. Additionally, CD38 has been shown to be a prognostic marker for some leukemias and other diseases.
Type: Primary
Antigen: CD38
Clonality: Monoclonal
Clone: HIT2
Conjugation: PerCP-Cyanine5.5
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human