Universal multi-pole breaker lockout
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
066321 066320
BRDY066321EA 15.02 EUR
BRDY066321 BRDY066320
Universal multi-pole breaker lockout
Veiligheidssloten
This universal breaker lockout is compatible with most two or three pole breakers with a tie-bar, such as GE ITE, Sylvania, Challenger, Bryant, Westinghouse and General Switch Company.
  • Made of moulded glass-filled nylon
Use thumbscrews to clamp lockout securely over tie-bar, then insert lock to prevent clamp from being loosened.
Max. shackle diameter: 7,00 mm
Min. shackle clearance: 20,00 mm