BrightVision+ Anti-IgG (Poly-AP)

Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

C-DPVB110AP C-DPVB500AP C-DPVB55AP C-DPVB999AP
C-DPVB110APEA 742 EUR
C-DPVB110AP C-DPVB500AP C-DPVB55AP C-DPVB999AP
BrightVision+ Anti-IgG (Poly-AP)
Antilichamen
BrightVision+ histostaining reagents utilise a novel controlled polymerisation technology to prepare polymeric APlinker antibody conjugates. Comparing to conventional biotin-streptavidin based detection kits, BrightVision+ histostaining kits have the advantages of higher amplification power, biotin free and more consistent immunostaining outcomes on archival tissues and on difficult-to-work antibodies.

  • High quality secondary antibody for IHC applications, AP labeled
  • Antigen Specificity: Anti-Mouse IgG (H+L), Anti-Rabbit IgG (H+L), Anti Rat IgG (H+L)
  • Kit conatins post-antibody blocking (ready-to-use) Gold and poly-AP-anti-mouse/rabbit IgG (ready-to-use) Ruby

Biotin-free
Ready-to-use

Certificaten: Meets CE/IVD standards.

Bestelinformatie: Packages above 110 ml contain small empty bottles so users can create smaller working volumes and prevent possible contamination of the whole lot.

Leveringsinformatie: Shipped with blue ice. Store between 2 to 8 °C.

Opgelet: Irritating to eyes, respiratory systems and skin.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR