BrightVision+ Anti-IgG (Poly-HRP)
Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
C-DPVB110HRP C-DPVB500HRP C-DPVB55HRP C-DPVB999HRP
C-DPVB110HRPEA 866 EUR
C-DPVB110HRP C-DPVB500HRP C-DPVB55HRP C-DPVB999HRP
BrightVision+ Anti-IgG (Poly-HRP)
Antilichamen
BrightVision+ histostaining reagents utilise a novel controlled polymerisation technology to prepare polymeric HRP-linker antibody conjugates. Comparing to conventional biotin-streptavidin based detection kits, BrightVision+ histostaining kits have the advantages of higher amplification power, biotin free and more consistent immunostaining outcomes on archival tissues and on difficult-to-work antibodies.
  • High quality secondary antibody for IHC applications, HRP labelled
  • Antigen Specificity: Anti-Mouse IgG (H+L), Anti-Rabbit IgG (H+L)
  • Kit contains post-
More Product Information