BrightVision+ Anti-IgG (Poly-AP)
Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
DPVR110AP DPVR55AP DPVR500AP DPVR999AP
DPVR110APEA 905 EUR
DPVR110AP DPVR55AP DPVR500AP DPVR999AP
BrightVision+ Anti-IgG (Poly-AP)
Antilichamen
BrightVision histostaining reagents utilise a novel controlled polymerisation technology to prepare polymeric APlinker antibody conjugates. Comparing to conventional biotin-streptavidin based detection kits, BrightVision histostaining kits have the advantages of higher amplification power, biotin free and more consistent immunostaining outcomes on archival tissues and on difficult-to-work antibodies.
  • High-quality secondary antibody for IHC applications, AP labelled
  • Antigen specificity: Anti-rabbit IgG (H+L)
Biotin-free
Ready-to-use
Certificaten: Meets CE/IVD standards.
Bestelinformatie: Packages above 110 ml contain small empty bottles so users can create smaller working volumes and prevent possible contamination of the whole lot.
Leveringsinformatie: Shipped with blue ice. Store between 2 to 8 °C.
Opgelet: Irritating to eyes, respiratory systems and skin.