BrightVision+ Anti-IgG (Poly-HRP)

Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

DPVO110HRP DPVO500HRP DPVO55HRP DPVO999HRP
DPVO110HRPEA 1250 EUR
DPVO110HRP DPVO500HRP DPVO55HRP DPVO999HRP
BrightVision+ Anti-IgG (Poly-HRP)
Antilichamen
BrightVision histostaining reagents utilise a novel controlled polymerisation technology to prepare polymeric HRP linker antibody conjugates. Comparing to conventional biotin-streptavidin based detection kits, these kits have the advantages of higher amplification power, biotin free and more consistent immunostaining outcomes on archival tissues and on difficult-to-work antibodies.

  • High-quality secondary antibody for IHC applications, HRP labelled
  • Antigen Specificity: Anti-Mouse IgG (H+L), Anti-Rabbit IgG (H+L), Anti-Rat IgG (H+L)

Kit contains Poly-More Product Information
Order Now

Specificaties

antigeen symbool IgG (H+L)
Environmentally Preferable
Type antilichaam Ondergeschikt
Conjugatie Poly-HRP (mierikswortelperoxidase)
Reactivity Rabbit,Mouse
Host
Isotype
ImmunoChemie Yes
Opslagtemperatuur +2 °C to +8 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR