MP-AES torches
Leverancier: Agilent
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
G8000-64140 G8000-70002 G8003-70000 G8003-20011 G8003-60018 G8003-68003
HEWLG8000-64140EA 74 EUR
HEWLG8000-64140 HEWLG8000-70002 HEWLG8003-70000 HEWLG8003-20011 HEWLG8003-60018 HEWLG8003-68003
MP-AES torches
Benodigdheden voor chromatografie Chromatografie reserveonderdelen
MP-AES Torches for the 4100/4200 Microwave Plasma-Atomic Emission Spectrometer.
  • Accuracy even with difficult samples — the vertical plasma torch provides excellent performance for tough samples with axial viewing for the best sensitivity
  • Reduce sample preparation
  • The axially viewed vertical plasma handles your toughest samples — from food and soil digests to soil extractions
  • Improve productivity when running hydrofluoric (HF) acid digests
  • The inert torch enables direct analysis of HF acid digests directly without a prior neutralization step