pH and ORP combination electrodes, TopLine
Leverancier: SI Analytics
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
285111190 285111195 285111200 285111205 285111210
662-2110EA 233 EUR
662-2110 662-2111 662-2112 662-2113 662-2115
pH and ORP combination electrodes, TopLine
Elektroden pH-elektroden
TopLine electrodes combine reliable measuring results with a maximum dwell time. They were developed for a wide range of applications in the lab, field and processing of wastewater via emulsions, solutions with proteins, suspensions all the way to purely aqueous samples.
  • Peek shaft for the highest mechanical and chemical resistance
  • Most precise measurements thanks to proven process membrane glasses and reference system
  • Maximum dwell time by means of a reference electrode with microdispersed KCI solid electrodes (DuraLid) as well as a multi-diaphragm system with external fibre wick hole and internal labyrinth reference element (LoopRef)
  • Available as analogue electrodes or as digital electrodes with IDS® and Memosens® head
pH range: 0 - 14
Zero point [pH]: 7
Electrolyte: Duralid polymer electrolyte with oversaturated dispersed KCl
Outer diaphragm: Fiber packing hole
Membrane: A-glass; H-glass (TopLine 80-120 pH, TopLine 81-120 pHT VP, TopLine 83-120 NMSN); Pt (ORP electrodes)
Reference system: LoopRef Labyrinth
Response time (98% between pH 4...7): < 20 sec.