ESD safety shoes, lace-up, uvex 1, 8546
Leverancier: UVEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8546.8/35 8546.8/36 8546.8/37 8546.8/38 8546.8/39 8546.8/40 8546.8/41 8546.8/42 8546.8/43 8546.8/44 8546.8/45 8546.8/46 8546.8/47 8546.8/48 8546.8/49 8546.8/50 8546.8/51 8546.8/52 8546.9/35 8546.9/36 8546.9/37 8546.9/38 8546.9/39 8546.9/40 8546.9/41 8546.9/42 8546.9/43 8546.9/44 8546.9/45 8546.9/46 8546.9/47 8546.9/48 8546.9/49 8546.9/50 8546.9/51 8546.9/52
UVEX8546.8/35EA 133 EUR
UVEX8546.8/35 UVEX8546.8/36 UVEX8546.8/37 UVEX8546.8/38 UVEX8546.8/39 UVEX8546.8/40 UVEX8546.8/41 UVEX8546.8/42 UVEX8546.8/43 UVEX8546.8/44 UVEX8546.8/45 UVEX8546.8/46 UVEX8546.8/47 UVEX8546.8/48 UVEX8546.8/49 UVEX8546.8/50 UVEX8546.8/51 UVEX8546.8/52 UVEX8546.9/35 UVEX8546.9/36 UVEX8546.9/37 UVEX8546.9/38 UVEX8546.9/39 UVEX8546.9/40 UVEX8546.9/41 UVEX8546.9/42 UVEX8546.9/43 UVEX8546.9/44 UVEX8546.9/45 UVEX8546.9/46 UVEX8546.9/47 UVEX8546.9/48 UVEX8546.9/49 UVEX8546.9/50 UVEX8546.9/51 UVEX8546.9/52
ESD safety shoes, lace-up, uvex 1, 8546
Schoenen en laarzen
Metal-free, extremely lightweight, flexible, multifunctional safety shoe made from breathable perforated high-tech micro velour upper, soft padded tongue and collar, plus a dual-density PUR sole (oil and petrol resistant, anti-static), free of lacquer-wetting inhibitors such as silicones or phthalates.
  • Climazone technology for improved comfort
  • Breathable distance mesh lining
  • Removable anti-static comfort insole with moisture control system; orthopaedic insoles possible
  • Anti-slip sole with shock absorption at heel and front of the foot
  • Virtually seamless design eliminates potential pressure points and blisters
  • Elastic laces for quick and individual adjustment
  • 100% metal-free xenova® toecap, non magnetic (200 J)
  • Fulfils ESD requirements: Resistance to ground inferior to 35 megaohms
Suitable for people allergic to chrome.
Colour: Black/red
Certificaten: EN 61340 (ESD); DGUV112-191; EN ISO 20345:2011; S1, SRC