Closures for headspace vials
Leverancier: PerkinElmer
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
B0104242 B0104243 B4000022 B4000025 B0104244 B0104241 N9303991 N9303992 N6356559 N6356566 N9302975 N9302978
PERCB0104242EA 1200 EUR
PERCB0104242 PERCB0104243 PERCB4000022 PERCB4000025 PERCB0104244 PERCB0104241 PERCN9303991 PERCN9303992 PERCN6356559 PERCN6356566 PERCN9302975 PERCN9302978
Closures for headspace vials
Sluitingen Sluitingen voor chromatografievials
Caps for 20 ml glass headspace crimp top round bottom vials which have a 23 mm OD and 75,5 mm height.