Sampling kits, SteriPlast
Leverancier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5378-8003 5378-8009
231-0591EA 37.1 EUR
231-0591 231-0592
Sampling kits, SteriPlast
Scheppen
The sampling set SteriPlast Kit consists of ten sample spatulas or ten sample scoops and ten SteriBag Premium tamper-proof sample bags. Thus the kit is the ideal combination for sampling where purity and sterility is essential.
  • All parts are sterilised by gamma rays
  • Suitable for use with foodstuffs
  • Scoops or spatulas are cleanroom manufactured and individually packed
  • Sample bag SteriBag Premium has an integrated sealing and is liquid-tight
  • Unique tracking code on both the bag and the tear-off field guarantees complete documentation and traceability
Leveringsinformatie: SteriPlast kit includes ten 50 ml scoops or ten spatulas (150 mm insertion depth) and ten 300 ml SteriBag Premium sample bags.