Wire basket for laboratory glassware
Leverancier: WDF - Funke GmbH & Co. KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
520-08-11 520-05-15 520-08-15 520-13-15 520-20-13 520-20-15
216-7236EA 0 EUR
216-7236 216-7244 216-7245 216-7246 WCDF520-20-13 WCDF520-20-15
Wire basket for laboratory glassware
Manden Draadmanden
Made of wire.
  • Spot-welded grid 8×8×1 mm
  • Good chemical resistance
  • Good temperature resistance
These wire baskets are designed for laboratory glassware.