GLP-kits, F1-Clip Tip™
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
4701140N 4701150N
613-5538EA 0 EUR
613-5538 613-5539
GLP-kits, F1-Clip Tip™
Pipetten Micropipetten
Support good laboratory pipetting in your lab with the Thermo Scientific™ F1-ClipTip™ GLP Kits, which offer a variety of single channel pipettes or a mix of single channels and a multichannel pipette packaged with compatible ClipTip tips, and a convenient pipette stand.
  • Worry-free pipetting is finally here with the only tip that clips
  • Sealed in security of ClipTip Technology. Feel the difference
  • Eliminate the frustration of banging tips on your pipette with a light touch, each tip is clipped securely on the pipette
  • Reduce the risk of loose or leaking tips when ClipTip pipette tips are on, they are sealed and will not loosen
  • The ClipTip pipetting system remains sealed until you decide to eject tips, even in the most comprehensive applications, including multiple tip touches and mixing cycles
  • Enable consistent and reproducible pipetting from user to user higher-quality results and more efficient research Ergonomically in-touch Lightweight handle design combined with low plunger, tip attachment and ejection forces help reduce the risk of RSI Silent, effortless volume adjustment enjoy the silent and lighter volume adjustment mechanism with over 50% lower forces compared to earlier models
  • Secure volume lock The secure volume setting prevents undesirable volume drift during operation
  • Simply lift the plunger up to set volume and press down to lock in place with minimum effort
  • Adjustable comfort the 120° adjustable finger rest - for both right- and left-handed operator comfort - allows the hand to relax during pipetting cycles
  • Super blow-out 150% increase in air boost to ensure efficient delivery of micro-volumes and prevent capillary action in 50 µl models and below
  • Wide Selection of ClipTip Pipette Tips Choose from seven sizes in 96-format: 12,5 Ext, 20, 50, 200, 300, 300 Ext,1000 µl
Select the kit that is right for you, and take advantage of the F1-ClipTip™ performance and lightweight design to maximise your comfort.
Lifetime warranty with convenient online registration, and may vary by country.
Bestelinformatie: Kit 1 includes: Three Thermo Scientific & trade F1-ClipTip pipettes from 1 to 1000 µl
- 1 to 10 µl
- 10 to 100 µl
- 100 to 1000 µl
- Grip Strip for each handle to improve grip
- ClipTip tips: 12,5 Ext: 1×96 200: 1×96 1000: 1×96-F-stand-Good Laboratory Pipetting Guide-Reagent reservoir demo pack
Kit 2 includes: Four Thermo Scientific & trade F1-ClipTip pipettes from 2 to 1000 µl
- 2 to 20 µl
- 20 to 200 µl
- 100 to 1000 µl
- 8-ch 30 to 300 µl
- Grip Strip for each handle to improve grip
- ClipTip tips: 20:1×96 200: 1×96 300: 1×96 1000: 1×96
- F-stand
- Good Laboratory Pipetting Guide
- Reagent reservoir demo pack