Calibration standards for thickness gauges
Leverancier: Industrial Physics
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
LD5406 LD5404 LD5403 LD5401 LD5402 LD5405
649-0025EA 559 EUR
649-0025 649-0023 649-0022 649-0020 649-0021 649-0024
Calibration standards for thickness gauges
Coatinglaag testers
These coated metal calibration standards consist of a set of four calibration plates.
They are composed of an epoxy coating on steel or aluminium when many gauges are in use or when accuracy verification is performed frequently.
Certificaten: ISO/QS-9000 and in-house quality control requirements.