PIG® Carry Bags for DrainBlocker® Drain Cover
Leverancier: NEW PIG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PLR232-36IN PLR232-24IN PLR232-48IN
121-0549EA 96.9 EUR
121-0549 121-0548 NEWPPLR232-48IN
PIG® Carry Bags for DrainBlocker® Drain Cover
Absorptiematerialen en bescherming tegen morsen
When a spill strikes, every second counts. This carry bag makes it easy to locate your drain cover and get there fast.
  • Wall-mountable drain seal bag has "J" hooks and "D" rings (included) for easy installation
  • Made of weather-resistant, PVC-supported nylon; protects Drain Cover and withstands damp environments
  • Yellow colour is highly visible for easy location during a spill
Designed to hold square DrainBlocker Drain Covers for easy transportation and fast spill response.