Scoops for foodstuffs, SteriPlast®
Leverancier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5378-3007 5378-3013 5378-3015 5378-3001 5378-3003 5378-3005 5378-3019 5378-3020
231-0585EA 22.2 EUR
231-0585 231-0589 231-0590 231-0582 231-0583 231-0584 231-0598 331-0366
Scoops for foodstuffs, SteriPlast®
Scheppen
The SteriPlast® food scoop, made of blue PS, is produced and assembled in a class 7 cleanroom (10000, except for the 1000 ml scoop), individually packaged for disposable use and then sterilised by gamma rays. The food scoop is ideal for powders, granulates and pastes.
  • Can be used as part of HACCP/IFS foreign object management
  • Especially for the food, animal feed and pharmaceutical industries
  • Individually packaged and sterilised by gamma rays
  • Production, assembly and packaging according to clean room class 7
  • Complies with EU food requirements and FDA