Magnet probes for curvex datalogger
Leverancier: Industrial Physics
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CX2062 CX2060 CX2050
SIMECX2062EA 230 EUR
SIMECX2062 SIMECX2060 SIMECX2050
Magnet probes for curvex datalogger
Temperatuursondes
Magnet probes to measure object temperature, fitted with an ultra strong magnet, but still with a very low mass and size.
  • Magnet Ø 14 mm
  • Energy and/or water efficient
  • Recycled/renewable content
The actual sensor is thermally isolated from the magnet in order not to affect the part's temperature. This sensor is suited for use on round parts, such as tubes.