Spatulas for foodstuffs, SteriPlast®
Leverancier: Bürkle
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SteriPlast®
231-0586EA 18.3 EUR
231-0586
Spatulas for foodstuffs, SteriPlast®
Spatels
The SteriPlast® spatula for foodstuffs, made of PS, is produced and assembled in a class 7 cleanroom (10,000), individually packaged for disposable use and then sterilised by gamma rays. The spatula is ideal for sampling powders, granulates and pastes.
  • Can be used as part of HACCP/IFS foreign object management
  • Especially for the food, animal feed and pharmaceutical industries
  • Individually packaged and sterilised by gamma rays
  • Production, assembly and packaging according to cleanroom class 7
  • Complies with EU food requirements and FDA regulations
The shade of blue that is used does not occur in a natural form in foodstuffs. Because of this, blue products or parts of them can be quickly found and easily identified in a visual check if they are ever mixed into production.