Surface probes, clamp-type, for Curvex datalogger
Leverancier: Industrial Physics
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CX2049 CX2048
SIMECX2049EA 322 EUR
SIMECX2049 SIMECX2048
Surface probes, clamp-type, for Curvex datalogger
Temperatuursondes
Clamp-type probes to measure object temperature (480 °C).
  • Max. object thickness is 25 mm