PIG Blue® absorbent mat pads and rolls
Leverancier: NEW PIG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BLU101 BLU103 BLU100
NEWPBLU101EA 154 EUR
NEWPBLU101 NEWPBLU103 NEWPBLU100
PIG Blue® absorbent mat pads and rolls
Absorptiematerialen en bescherming tegen morsen
PIG Blue is the most absorbent mat. Maximum absorbency soaks up more so you use less on big spills and high-volume leaks.
  • Consistent construction produces a mat that's free of light spots to absorb liquid evenly for better wicking and retention
  • FibreFusion Technology fuses superabsorbent natural fibres with strong synthetic fibres for one high-performance absorbent
  • Stays together, even when picking up or shifting a fully saturated mat
  • Contains 90% recycled fibres; ideal in situations where green products are desired or required
  • Fire retardant for added