PIG® Rip-&-Fit® universal mat
Leverancier: NEW PIG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MAT242 MAT243
121-0524EA 86.5 EUR
121-0524 NEWPMAT243
PIG® Rip-&-Fit® universal mat
Absorptiematerialen en bescherming tegen morsen
Extra perforations let you create custom sizes of PIG Mat without cutting. Just "rip" it along the easy-tear perfs and "fit" it into place.
  • Extra perforations in our Rip-&-Fit Mat allow you to easily "rip" this PIG Mat and "fit" it to your application without cutting
  • No matter how you rip it, this mat features the proven eight-layer, 100% PP construction of PIG Mat
  • Exclusive dimple pattern speeds wicking of liquid throughout mat for faster, easier cleanup
  • Highly absorbent, fine-fibre construction won't leave behind liquids or fibre residue
  • Absorbs and