Surface probes, clamp-type, for Curvex datalogger
Leverancier: Industrial Physics
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CX2040 CX2041 CX2030 CX2045
SIMECX2040EA 204 EUR
SIMECX2040 SIMECX2041 SIMECX2030 SIMECX2045
Surface probes, clamp-type, for Curvex datalogger
Temperatuursondes
Small and elegant clamp-type probes to measure temperature of any object.
  • Max. object thickness is 20 mm
  • Energy and/or water efficient
  • Recycled/renewable content
Silver tipped sensor is thermally isolated from the clamp by ceramics. The capton-wrapped, metal braid and PTFE coated probe cables last significantly longer than other designs.