Precast agarose IEF plates, IsoGel®
Leverancier: LONZA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
56015 56024 56018
LONZ56015EA 644 EUR
LONZ56015 LONZ56024 LONZ56018
Precast agarose IEF plates, IsoGel®
Gel voor elektroforese Voorgegoten gels
Precast gels for the analysis of antibodies and proteins up to 2000 kDa.
  • Fast and easy handling
  • Gel size of 125×100 mm fits most IEF chambers
Gel is supported on GelBond® PAG support film to provide dimensional stability throughout IEF processing.