Temperature probes
Leverancier: Industrial Physics
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
TE5103 TE5125 TE5110 TE5109 TE5108 TE5106 TE5105 TE5104
SIMETE5103EA 358 EUR
SIMETE5103 SIMETE5125 SIMETE5110 SIMETE5109 SIMETE5108 SIMETE5106 SIMETE5105 SIMETE5104
Temperature probes
Temperatuursondes
These temperature probes can be connected to instruments suited for changeable probes (type ‘K’) using a plug connection.
  • Provided with a firm hand-grip and a one meter curled cable
  • Thicker probes have a longer response time