Fluoride calibration standards for ISE measurement, Orion™
Leverancier: Thermo Orion
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
040906 940916 O40907 O40908 940906 940907
662-0682EA 175 EUR
662-0682 ORIO940916 662-0683 662-0684 662-0826 662-0827
Fluoride calibration standards for ISE measurement, Orion™
Standaarden Anionische standaarden Fluoride standaarden
Fluoride standards are pre-made with total ionic strength adjustor buffer (TISAB) and requiring the TISAB be added only to your samples.