Orion™ Gas-Sensing ISE Membranes
Leverancier: Thermo Orion
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
951204 950204 951205 951215
662-0856EA 265 EUR
662-0856 ORIO950204 662-1515 662-1671
Orion™ Gas-Sensing ISE Membranes
Elektroden ISE-elektroden
These fresh membranes provide optimum performance with gas-sensing ion selective electrodes. Ammonia electrodes require different membranes if using standard electrode or high performance ammonia electrode.
  • Loose or preassembled membranes
  • Easy to use