HPLC columns, Polaris™ C8-ether
Leverancier: VARIAN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A2030050X020 A2030150X046 A2031050X020 A2031050X046 A2031150X020 A2031250X046 A2030100X020 A2030100X030 A2030100X046 A2030150X020 A2030250X020 A2030250X030 A2030250X040 A2030250X100 A2031050X030 A2031100X020 A2031100X046 A2031250X020
VARIA2030050X020EA 614 EUR
VARIA2030050X020 VARIA2030150X046 VARIA2031050X020 VARIA2031050X046 VARIA2031150X020 VARIA2031250X046 VARIA2030100X020 VARIA2030100X030 VARIA2030100X046 VARIA2030150X020 VARIA2030250X020 VARIA2030250X030 VARIA2030250X040 VARIA2030250X100 VARIA2031050X030 VARIA2031100X020 VARIA2031100X046 VARIA2031250X020
HPLC columns, Polaris™ C8-ether
Kolommen voor chromatografie
These Polaris™ phases have rapid high resolution separations based on ultra pure silica to which alkyl chains and polar modifications are attached.
  • Enhanced retention and peak shape of polar analytes
  • Additional selectivity for H-bond donors
  • High aqueous compatibility
  • No phase collapse