Guard columns, MetaGuard®
Leverancier: VARIAN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
A2000MG A2000MG2 A2001MG A2001MG2 A2030MG A2031MG2 A3000MG A3000MG1 A3000MG2 A3001MG A3001MG2 A3002MG A3002MG2 A2001MG1 A2002MG A2005MG A2010MG A2011MG A3031MG2 A3031MG A3030MG A3030MG2 A3040MG A3040MG1 A3040MG2 A3041MG A3041MG2 A3041MG1
VARIA2000MGEA 325 EUR
VARIA2000MG VARIA2000MG2 VARIA2001MG VARIA2001MG2 VARIA2030MG VARIA2031MG2 VARIA3000MG VARIA3000MG1 VARIA3000MG2 VARIA3001MG VARIA3001MG2 VARIA3002MG VARIA3002MG2 VARIA2001MG1 VARIA2002MG VARIA2005MG VARIA2010MG VARIA2011MG VARIA3031MG2 VARIA3031MG VARIA3030MG VARIA3030MG2 VARIA3040MG VARIA3040MG1 VARIA3040MG2 VARIA3041MG VARIA3041MG2 VARIA3041MG1
Guard columns, MetaGuard®
Kolommen voor chromatografie
These MetaGuard columns are stand-alone, disposable guard columns that easily connect directly into the end fitting analytical columns.
  • Fingertight connection with wide range of phases available
  • No appreciable effect on column efficiency or peak shape
  • Can be used in column heating applications where the space consideration is crucial
Forms a fingertight, very low dead-volume connection, which minimizes sample dispersion, maximizes system efficiency, and is leak-free to over 345 bar.