n-Dodecyl acrylate 90% stabilised, Technical Grade
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
Acrylic acid Lauryl ester, Acrylic acid Dodecyl ester, Dodecyl Acrylate, Lauryl Acrylate
Stabilisatie:
60 - 100 ppm Stabilised with 4-methoxyphenol (MEHQ)
H60625.AE
H60625.AP
H60625.AEEA
54.9
EUR
H60625.AE
H60625.AP
n-Dodecyl acrylate 90% stabilised, Technical Grade
n-Dodecyl acrylate
Formule:
C₁₅H₂₈O₂ MW: 240.387 g/mol Kookpunt: 120 °C (0.75 mmHg) Dichtheid: 0.884 Vlampunt: 110 °C (230 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00042871 CAS nummer: 2156-97-0 EINECS: 218-463-4 UN: 3082 ADR: 9 III |
Specification Test Results
Synonyms | Lauryl acrylate, Acrylic acid n-dodecyl ester |
Assay (HPLC) |
|
Refractive Index | 1,4450 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...