Soy hydrolysate enzymatic, powder, MP Biomedicals
Leverancier: MP Biomedicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0210294105 0210294101
ICNA0210294105EA 0 EUR
ICNA0210294105 ICNA0210294101
Soy hydrolysate enzymatic, powder, MP Biomedicals
Media voor microbiologie Additieven en supplementen voor microbiologie
Soy hydrolysate is an enzymatic (papain) digest of soybean meal. It is a plant source peptone. Soy hydrolysate is useful in media where high clarity is not required.
  • Total Nitrogen- ≥8,0%
  • Nitrogen, AN/TN - 17,0 - 28,0
  • Std. Plate Count ≤10 000
  • Enterobacteriaceae ≤10 CFU/G
Soluble in 100 mg/ml water.