2-Fenoxyethanol
Leverancier: MP Biomedicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: ω-Hydroxyphenetole, β-Hydroxyphenetole, Phenyl cellosolve, Phenoxyethanol, Ethylene glycol monophenyl ether
0215014880 0215014890
ICNA0215014880EA 42.9 EUR
ICNA0215014880 ICNA0215014890
2-Fenoxyethanol
2-Fenoxyethanol
Important notice: this product belongs to MP Biomedicals rare chemical library. Most rare chemical products are available for immediate delivery and sold "As Is". MP biomedicals does not collect analytical data for those products. All sales are final. However, analytical test may be performed upon custom request for bulk orders.
Opgelet: Research Use Only (RUO).
Formule: C₆H₅OCH₂CH₂OH
MW: 138.17 g/mol
Kookpunt: 245
Smeltpunt: 57 ° F (literatuur)
Vlampunt: 121
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002857
CAS nummer: 122-99-6
Merck Index: 12,07410
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d27cc4d39b806528798164961c23a1285d41d6a9&vid=a46b721cc69101d0c50dc49d9687445b395a3766&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Identity Test Passes
FTIR Conforms to Standard