Cinnamaldehyde ≥99%, liquid
Leverancier: MP Biomedicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: trans-Cinnamaldehyde, 3-Phenylacrylaldehyde
0215069080 0215069090
ICNA0215069080EA 85.8 EUR
ICNA0215069080 ICNA0215069090
Cinnamaldehyde ≥99%, liquid
Cinnamaldehyde
FEMA Number 2286, Flavis number 5.014 , Council of Europe no. 102c, Organoleptic: cinnamon; clove; spicy, allergen: no known allergens.
  • Presentation: Liquid
  • Vapor Pressure: 2.25 hPa at 20 °C (Lit.)
  • Refractive Index: n20/D 1.621(Lit.)
  • Solubility: Misc with alcohol, ether, chloroform, oils.
  • Storage temperature: +4°C, store under nitrogen
Formule: C₉H₈O
MW: 132.16 g/mol
Kookpunt: 246 °C (1014 hPa) (literatuur)
Smeltpunt: –7.5 °C (literatuur)
Dichtheid: 1.0497 g/cm³ (20 °C) (literatuur)
Vlampunt: 138
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00007000
CAS nummer: 104-55-2
Merck Index: 12,02356
Order Now
Specification Test Results
Identity Test Passes
Purity ≥99%
FTIR Conforms to Reference