DL-Adrenaline, tan powder

Leverancier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

0215106425 0215106480
ICNA0215106425EA 320 EUR
ICNA0215106425 ICNA0215106480
DL-Adrenaline, tan powder
DL-Adrenaline

(+/-)-Epinephrine is also known as adrenaline and is a hormone as well as a neurotransmitter that is part of a group of monoamines called catecholamines.


It is used medically to treat a number of cardiac and respiratory conditions as well as serving as a local anesthetic.
(±)-Epinephrine is an adrenoceptor agonist. The L-form is a direct-acting sympathomimetic agent with pronounced effect on alpha- and beta-adrenergic receptors. Its action is likened to endogenous adrenaline released from the adrenal medulla in response to extreme physiological conditions.


Soluble in 1 M HCI (50 mg/ml); freely soluble in water; sparingly soluble in ethanol (95% v/v)


Opgelet: Research Use Only (RUO).

Formule: C₉H₁₃NO₃
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00063027
CAS nummer: 329-65-7
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Standard


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR