Alphacel Non-Nutritive Bulk, MP Biomedicals
Leverancier: MP Biomedicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0290045305 0290045325 0290045380
ICNA0290045305EA 66.6 EUR
ICNA0290045305 ICNA0290045325 ICNA0290045380
Alphacel Non-Nutritive Bulk, MP Biomedicals
Dierenbenodigdheden Voeding voor dieren
An extremely fine, white, odorless tasteless, water insoluble, chemically pure cellulose powder, manufactured from natural fibres.
  • Presentation: White Powder
  • Store at room temperature (15 to 30 °C)
Alphacel Non-Nutritive Bulk is used to study the effect of conjugated linoleic acids from beef or industrial hydrogenation on growth and adipose tissue characteristics of rats. It is also used as a harmless, biologically acceptable, bulking agent in the specialised diets of laboratory animals.
Alphacel is manufactured by a dry, mechanical process from bleached and purified wood pulp.
An extremely fine, white, odorless tasteless, water insoluble, chemically pure cellulose powder, manufactured from natural fibers.
Alphacel non-nutritive bulk is used to study the effect of conjugated linoleic acids from beef or industrial hydrogenation on growth and adipose tissue characteristics of rats. It is also used as a harmless, biologically acceptable, bulking agent in the specialised diets of laboratory animals.