Salt Mixture, U.S.P. XIII No.2, MP Biomedicals
Leverancier: MP Biomedicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0290284502 0290284510
ICNA0290284502EA 123 EUR
ICNA0290284502 ICNA0290284510
Salt Mixture, U.S.P. XIII No.2, MP Biomedicals
Dierenbenodigdheden Voeding voor dieren
MP recommends the use of supplement U.S.P. XIII, No. 2 Mineral Mix (cat # 960300) in conjunction with this formulation for long term growth and development of the rat.
Components 10 kg
Monocalcium Phosphate 1362 g
Calcium Lactate 3276 gm
Ferric Citrate (16 to 17% Fe) 300 g
Magnesium Sulfate 1376 gm
Potassium Phosphate Dibasic 2400 g
Monosodium phosphate 876 g
Sodium Chloride 438 g