Natural Corn Granules, MP Biomedicals
Leverancier: MP Biomedicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0290141125 0290141180
ICNA0290141125EA 85.4 EUR
ICNA0290141125 ICNA0290141180
Natural Corn Granules, MP Biomedicals
Dierenbenodigdheden Voeding voor dieren
Corn is used as pet nutrition and natural pet food.
It contains carbohydrate (77%), protein (8%), fat (2%), fiber (2%) and water (11%).
Opgelet: Store at ambient temperature.