Methyl palmitoleate, liquid

Leverancier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: Palmitoleic acid methyl ester, (Z)-Methyl hexadec-9-enoate

0215604890 0215604891
ICNA0215604890EA 198 EUR
ICNA0215604890 ICNA0215604891
Methyl palmitoleate, liquid
Methyl palmitoleate
Formule: C₁₇H₃₂O₂
MDL Nummer: MFCD00042911
CAS nummer: 1120-25-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR