Ficoll 400, White powder, MP Biomedicals
Leverancier: MP Biomedicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0215632810 0215632825 0215632880
ICNA0215632810EA 39.8 EUR
ICNA0215632810 ICNA0215632825 ICNA0215632880
Ficoll 400, White powder, MP Biomedicals
Media voor celscheiding
Polysucrose is water-soluble, nontoxic, resistant to intestinal enzymes, spherical and can be produced with a molecular weight distribution that relates to the size of many normal food proteins. It is a nonionic synthetic polymer of sucrose.
  • MW: 400000 g/mol
Polysucrose 400 is used for cell separation and organelle isolation. It has been used in a study to develop a new method for the purification of human motile spermatozoa. It has also been used in a study that investigated the antiproliferative and antitumor activities of D-reverse peptides.
Store at Room Temperature (15 - 30 °C)
Soluble in 100 mg/ml Water.