Ijzer(II)sulfaat heptahydraat ≥99%, bluish green granular powder
Leverancier: MP Biomedicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Iron vitriol, Ferrous sulphate heptahydrate
0215804883 0215804891
ICNA0215804883EA 55.9 EUR
ICNA0215804883 ICNA0215804891
Ijzer(II)sulfaat heptahydraat ≥99%, bluish green granular powder
Ijzer(II)sulfaat heptahydraat
Ferrous sulphate heptahydrate used in the manufacturing of iron compounds, in electroplating baths, aluminium etching, process engraving and lithography, and in redox polymerisation. It is used as a reducing agent in chemical processes, leather dyes and writing inks.
Also, it is a component of weed killers, wood preservatives and water treatment processes. It is used in prevention of chlorosis in plants and therapeutically as hematinic. In veterinary medicine, it is used as an astringent to combat iron deficiency.
Soluble in water (100 mg/ml); practically insoluble in alcohol.
Formule: FeSO₄·7H₂O
MW: 278.02 g/mol
Smeltpunt: 64
MDL Nummer: MFCD00149719
CAS nummer: 7782-63-0
Merck Index: 13,04091
Order Now
Specification Test Results
Identity Test Passes
Purity ≥99%
FTIR Conforms to Reference