Ursodeoxycholic acid ≥99%, white powder
Leverancier: MP Biomedicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0215825201
0215825205
0215825225
ICNA0215825201EA
57.9
EUR
ICNA0215825201
ICNA0215825205
ICNA0215825225
Ursodeoxycholic acid ≥99%, white powder
Ursodeoxycholic acid
Ursodeoxycholic acid (UDCS) is a cell protectant used extensively to mitigate hepatic and biliary diseases. Ursodeoxycholic acid may be used to study its specific activities that range from reduction of cholesterol absorpition, cholesterol gallstone dissolution to suppression of immune response.
Soluble in ethanol (50 mg/ml, clear to very slightly hazy).
Formule:
C₂₄H₄₀O₄ MW: 392.58 g/mol Kookpunt: 547 Smeltpunt: 203…204 °C (literatuur) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00003680 CAS nummer: 128-13-2 Merck Index: 12,10026 |
Specification Test Results
Identity Test | Passes |
Purity | ≥99% |
FTIR | Conforms to Standard |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...