Clomiphene citrate salt, off-white powder

Leverancier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

0219017401 0219017425 0219017405 0219017410
ICNA0219017401EA 28.5 EUR
ICNA0219017401 ICNA0219017425 ICNA0219017405 ICNA0219017410
Clomiphene citrate salt, off-white powder
Clomiphene citrate salt

Clomiphene Citrate Salt is a mixture of cis- and trans- isomers. Clomiphene citrate is a selective estrogen receptor modulator.


Freely soluble in methanol; slightly soluble in water and chloroform; sparingly soluble in ethanol (96%); practically insoluble in ether.

Formule: C₃₂H₃₆ClNO₈
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00058322
CAS nummer: 50-41-9

Order Now

Specification Test Results

Identity Test Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR