Amylopectin (from corn), white powder
Leverancier: MP Biomedicals
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0219504891
0219504883
ICNA0219504891EA
185
EUR
ICNA0219504891
ICNA0219504883
Amylopectin (from corn), white powder
Amylopectin
Amylopectin is a glucose-based, highly-branched polysaccharide which together with amylose makes starch. The glucosidic bonds of amylopectin are α-1,4 in the glucan chain and α-1,6 at the branch points.
The measurement of the amylopectin content of starches is an important quality parameter for starch processing. It is used in the study of the development of starches.
Formule:
C₃₀H₅₂O₂₆ Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00130510 CAS nummer: 9037-22-3 |
Specification Test Results
Form | White Powder |
Storage | Store at Room Temperature (15 - 30 °C) |
Source | Corn |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...