Hubbel, Mendel and Wakeman Salt Mixture
Leverancier: MP Biomedicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ICNA0290283802EA 76.7 EUR
ICNA0290283802
Hubbel, Mendel and Wakeman Salt Mixture
Dierenbenodigdheden Voeding voor dieren
Hubbel, Mendel and Wakeman powder does not contain zinc, is low in manganese and phosphorus, and has a high calcium to phosphorus ratio.
Designed for use in formulating animal research diets, especially with relatively high casein levels.
For complete composition see product datasheet.