Vitamin Diet Fortification Powder
Leverancier: MP Biomedicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ICNA0290465401EA 267 EUR
ICNA0290465401
Vitamin Diet Fortification Powder
Dierenbenodigdheden Voeding voor dieren
Vitamin diet fortification mixture formulation provides sufficient vitamin levels to prevent deficiencies in all animal species, giving precise control of vitamin levels.
The ingredients of the vitamin diet fortification mixture are triturated in dextrose (1 kg is sufficient for 100 lbs of diet).